  • Search Results
  • Effects of ultrafine petrol exhaust particles on cytotoxicity, oxidative stress generation, DNA damage and inflammation in human A549 lung cells and murine RAW 264.7 macrophages.
Effects of ultrafine petrol exhaust particles on cytotoxicity, oxidative stress generation, DNA damage and inflammation in human A549 lung cells and murine RAW 264.7 macrophages.
Environmental toxicology and pharmacology (2014-09-01)
Mohan Durga, Soundararajan Nathiya, Abbu Rajasekar, Thiyagarajan Devasena
ABSTRACT
Air pollution has persistently been the major cause of respiratory-related illness and death. Environmental pollutants such as diesel and petrol exhaust particles (PEPs) are the major contributors to urban air pollution. The aim of the present study was to characterize and investigate the in vitro cytotoxicity, oxidative stress, DNA damage and inflammation induced by PEPs. Cultured type II epithelium cells (human A549 lung cells) and alveolar macrophages (murine RAW 264.7 cells) were exposed to control, vehicle control and to different concentrations of PEPs for up to 24h. Each treatment was evaluated by cell viability, cytotoxicity, oxidative stress, DNA damage and inflammatory parameters. Overall in vitro studies demonstrated that both cell lines showed similar patterns in response to the above studies induced by petrol exhaust nanoparticles (PENPs). Vehicle control showed no changes compared with the control. In both cell lines, significant changes at the dose of 20 and 50μg/mL (A549 cell lines) and 10and 20μg/mL (macrophages) for PENPs were found. The reactive oxygen species production in both cell lines shot up in minutes, reached the maximum within an hour and came down after 4h. Hence, exposure to PENPs resulted in dose-dependent toxicity in cultured A549 cells and RAW 264.7 cells and was closely correlated to increased oxidative stress, DNA damage and inflammation.
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IL-6 sR ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material