  • Search Results
  • Loss of Dlg5 expression promotes the migration and invasion of prostate cancer cells via Girdin phosphorylation.
Loss of Dlg5 expression promotes the migration and invasion of prostate cancer cells via Girdin phosphorylation.
Oncogene (2014-03-26)
L Tomiyama, T Sezaki, M Matsuo, K Ueda, N Kioka
ABSTRACT
Dlg5 has been reported to participate in cancer progression; however, its role in prostate cancer still remains poorly understood. In this study, we demonstrate that Dlg5 is frequently downregulated in prostate cancer. We show here that Dlg5 is involved in the regulation of cell migration and cancer cell invasion. Knockdown of endogenous Dlg5 markedly increased prostate cancer cell migration and invasion. Our studies, for the first time, demonstrate the interaction between Dlg5 and Girdin, an actin-binding Akt substrate. Importantly, we found that levels of Akt-mediated Girdin phosphorylation (p-Girdin-Ser1416) are increased in Dlg5-depleted cells. Small interfering RNA directed against Girdin and wortmannin treatment, which was found to reduce Girdin phosphorylation, impaired the effect of Dlg5 depletion on cell migration. Taken together, our findings demonstrate that Dlg5 interacts with and inhibits the activity of Girdin, thereby suppressing the migration of prostate cancer cells.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Hematoxylin
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%