  • Search Results
  • Cellular uptake of coumarin-6 under microfluidic conditions into HCE-T cells from nanoscale formulations.
Cellular uptake of coumarin-6 under microfluidic conditions into HCE-T cells from nanoscale formulations.
Molecular pharmaceutics (2014-11-21)
S Pretor, J Bartels, T Lorenz, K Dahl, J H Finke, G Peterat, R Krull, A T Al-Halhouli, A Dietzel, S Büttgenbach, S Behrends, S Reichl, C C Müller-Goymann
ABSTRACT
In vitro studies of ocular bioavailability of active pharmaceutical ingredients (API) from colloidal drug delivery systems do not consider physiological shear stress generated by eyelid wiping and tear flow. The present study introduces a live cell imaging approach which enables the investigation of model drug uptake from various formulations under shear stress by using custom-made microchannels for the cultivation of human corneal epithelial cells (HCE-T). Coumarin-6 (C-6) was used as a model API incorporated into solid lipid nanoparticles and liposomes, and as an aqueous crystalline suspension. Confocal laser scanning microscopy visualized C-6 uptake into HCE-T cells in a time-resolved manner with an applied shear stress of 0.1 Pa. Static conditions were also studied for comparative purposes. Additionally, solid lipid nanoparticles (SLN) were labeled with a fluorescent phospholipid to check whether C-6 uptake was associated with SLN incorporation into the cells. Intact SLN were not incorporated into the cells, i.e., C-6 was passively redistributed from SLN to lipophilic cellular compartments. C-6 was enriched up to a given limit in HCE-T cells within 5 min of contact with the dispersions both under static and under flow conditions. The C-6 delivery rate from liposomes was superior to that from SLN whereby the suspension exhibited the lowest rate. C-6 release rates were comparable for static and flow conditions. Alternate flushing with formulations and buffer revealed that cells accumulated C-6. The results suggest that combining microfluidics with live cell imaging provides a valuable option for in vitro studies of ocular drug delivery.
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Calcein-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O