  • Search Results
  • AG4, a compound isolated from Radix Ardisiae Gigantifoliae, induces apoptosis in human nasopharyngeal cancer CNE cells through intrinsic and extrinsic apoptosis pathways.
AG4, a compound isolated from Radix Ardisiae Gigantifoliae, induces apoptosis in human nasopharyngeal cancer CNE cells through intrinsic and extrinsic apoptosis pathways.
Anti-cancer drugs (2014-12-19)
Xian-Zhe Dong, Ting-Ting Xie, Xiao-Jiang Zhou, Li-Hua Mu, Xiao-Li Zheng, Dai-Hong Guo, Ping Liu, Xiao-Yue Ge
ABSTRACT
3β-O-{α-L-Pyran rhamnose-(1→3)-[β-D-xylopyranose-(1→2)]-β-D-glucopyranose-(1→4)-[β-D-lucopyranose-(1→2)]-α-L-pyran arabinose}-cyclamiretin A (AG4) is a saponin component obtained from the Giantleaf Ardisia Rhizome (Rhizoma Ardisiae Gigantifoliae). The present study aimed to investigate the antitumor potential of AG4 and its possible mechanisms in human nasopharyngeal carcinoma cells (CNE). We exposed tumor cells to AG4 to investigate which cell line was the most sensitive to AG4. Cell viability was assessed using the MTT reduction assay, and the effects of AG4 on apoptosis, reactive oxygen species (ROS) content, mitochondrial membrane potential (MMP), and cell cycle were detected using a flow cytometer; the glutathione, superoxide dismutase and malondialdehyde activities were measured using colorimetric methods. The relative expressions of Bax, Bad, Bid, Bcl-2, and Fas mRNA were calculated using the (Equation is included in full-text article.)comparative method by real-time PCR studies and protein was detected by western blotting. AG4 markedly inhibited the growth of CNE cells by decreasing cell proliferation, inducing apoptosis, and blocking the cell cycle in the S phase. The release of caspase-3, caspase-8, and caspase-9 was stimulated by AG4 in CNE, and the decreased proliferation induced by AG4 was blocked by the inhibitor of pan caspase (Z-VAD-FMK). Moreover, the MMP was decreased in AG4-treated cells, and AG4-induced cell apoptosis was accompanied by a rapid and lasting increase in ROS, which was abolished by N-acetyl-L-cysteine (NAC); glutathione, superoxide dismutase, and malondialdehyde were regulated by AG4. AG4 inhibited Bcl-2 mRNA and protein expression and stimulated Bax, Bad, Bid, Fas mRNA, and protein expression in CNE cultures, suggesting an effect at the transcriptional and protein level. In addition, both the FasL inhibitor (AF-016) and the Bcl-2 family inhibitor (GX15-070) could prevent the cell apoptosis induced by AG4. The findings suggested that AG4-induced apoptosis in CNE cells involved a death receptor pathway and a Bcl-2 family-mediated mitochondrial signaling pathway by decreasing the MMPs in an ROS-dependent manner and regulating genes and proteins relative to apoptosis; also, regulation of cell cycles may also play a role in the antitumor mechanism of AG4.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%