  • Search Results
  • Plasma phosphatidylcholine alterations in cystic fibrosis patients: impaired metabolism and correlation with lung function and inflammation.
Plasma phosphatidylcholine alterations in cystic fibrosis patients: impaired metabolism and correlation with lung function and inflammation.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-03-21)
Judith Grothe, Joachim Riethmüller, Sandra M Tschürtz, Marco Raith, Chris J Pynn, Dieter Stoll, Wolfgang Bernhard
ABSTRACT
Liver impairment, ranging from steatosis to cirrhosis, is frequent in cystic fibrosis (CF) patients and is becoming increasingly significant due to their improved life expectancy. One aspect of hepatic alterations is caused by increased fecal loss of the essential nutrient choline, following enterohepatic bile phosphatidylcholine (PC) cycle impairment. Hepatic PC synthesis, both de novo and via phosphatidylethanolamine-N-methyl-transferase (PEMT), is essential for very low-density lipoprotein (VLDL) secretion. VLDL-PC in particular contributes to the organism's supply with polyunsaturated fatty acids (LC-PUFA), namely arachidonic (C20:4) and docosahexaenoic acid (C22:6). Consequently, choline deprivation and altered hepatic PC metabolism may affect plasma PC homeostasis and extrahepatic organ function. To investigate relationships between altered plasma choline and PC homeostasis and markers of lung function and inflammation in CF. To assess alterations in hepatic choline and PC metabolism of CF patients. Quantification of plasma/serum choline and PC species in adult CF patients compared to controls. Correlation of PC with forced expiratory vital capacity (FEV1) and interleukin 6 (IL-6) concentrations. Analysis of choline and PC metabolism in CF compared to controls, using deuterated choline ([D₉-methyl]-choline) labeling in vivo. Mean choline and PC concentrations in CF patients were lower than in controls. Choline and PC concentrations as well as fractions of C22:6-PC and C20:4-PC correlated directly with FEV1, but inversely with IL-6. Plasma concentrations of deuterated PC were decreased for both pathways, whereas only in PC synthesized via PEMT precursor enrichment was decreased. In CF patients, hepatic and plasma homeostasis of choline and PC correlate with lung function and inflammation. Impaired hepatic PC metabolism, exemplarily shown in three CF patients, provides an explanation for such correlations. Larger studies are required to understand the link between hepatic PC metabolism and overall clinical performance of CF patients, and the perspective of choline substitution of these patients.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis