  • Search Results
  • Evaluation of chemotherapeutics in a three-dimensional breast cancer model.
Evaluation of chemotherapeutics in a three-dimensional breast cancer model.
Journal of cancer research and clinical oncology (2015-03-17)
Carrie J Lovitt, Todd B Shelper, Vicky M Avery
ABSTRACT
Utilization of miniaturized three-dimensional (3D) cell culture-based assays enables investigation into the anticancer activity of drug candidates and further elucidation of the anticancer profile of standard-of-care chemotherapeutic agents against tumor cells. Drug discovery assays established using 3D cell culture, which better recapitulate the tumor microenvironment, may more accurately reflect the antitumor activity of compounds. Several standard-of-care anticancer drugs, epirubicin, paclitaxel and vinorelbine, were evaluated against a panel of breast cancer cell lines grown in a 3D cell culture microenvironment in the presence of extracellular matrix. A comparison of this antitumor activity in 3D conditions was made with that observed in traditional two-dimensional (2D) monolayer conditions. Examination of the above mentioned drugs against breast tumor cells cultured in 3D conditions demonstrated significantly altered potency and efficacy in comparison with cells propagated in a 2D monolayer system. The differences observed were cell line-dependent and drug-specific; the triple-negative cell line MDA-MB-231 and the endocrine receptor-positive cell line MCF-7 consistently displayed resistance to therapeutics with distinct modes of action (i.e., topoisomerase II and microtubules) in 3D cell culture in comparison with ErbB2 receptor-positive BT-474 cells. The data presented herein demonstrates the cellular viability and physical changes observed within the 3D spheroid following exposure to drug, which is not always reflected in 2D cell culture models.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥97%
Sigma-Aldrich
Resazurin sodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O