  • Search Results
  • Effects of phosphorus-restricted diet and phosphate-binding therapy on outcomes in patients with chronic kidney disease.
Effects of phosphorus-restricted diet and phosphate-binding therapy on outcomes in patients with chronic kidney disease.
Journal of nephrology (2014-03-07)
Domenico Russo, Antonio Bellasi, Andrea Pota, Luigi Russo, Biagio Di Iorio
ABSTRACT
Phosphorus is associated with mortality in patients with chronic kidney disease (CKD) not on dialysis, possibly through phosphorus-dependent vascular calcification. Although a phosphorus-restricted diet reduces serum phosphorus, it is unlikely that it reduces vascular calcification progression in CKD. This study evaluated whether a combined strategy of phosphorus-restricted diet and phosphate-binding therapy can reduce the risk of all-cause mortality and/or dialysis initiation by attenuating coronary artery calcification (CAC) progression in non-dialysis CKD patients. This was a post hoc analysis of a subgroup of patients from a study that evaluated the impact of two phosphorus binder regimens on hard outcomes in CKD. Patients (n = 113) with stage 3-4 CKD and evidence of CAC on a phosphorus-restricted diet were randomized to receive either calcium carbonate or sevelamer added to their phosphorus-restricted diet. End-points were death for any cause and initiation of dialysis. Patients were monitored to the first event or to conclusion of the 36-month follow-up. Overall, treatment with calcium carbonate was associated with increased CAC progression and occurrence of all-cause mortality, dialysis initiation, and the composite end-point. After adjustment for confounders, sevelamer use was the only independent predictive factor of reduced risk of each endpoint but only if CAC progression was either absent or moderate. Accelerated progression (annual CAC increase >75th percentile of the study cohort) increased the risk of all-cause mortality and composite end-point (p = 0.01) independently of the use of sevelamer. A significant reduction in all-cause mortality, dialysis initiation, and composite end-point risk was achieved by combining phosphorus-restricted diet and sevelamer in non-dialysis CKD patients with absent or moderate but not accelerated CAC progression. Future studies should investigate the role of serum phosphorus, the usefulness of a phosphorus-restricted diet, and the appropriateness of current normal ranges of serum phosphorus concentration in relation to events in non-dialyzed CKD patients.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Supelco
Calcium carbonate, certified reference material for titrimetry, certified by BAM, ≥99.5%
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard