  • Search Results
  • PLCE1 mRNA and protein expression and survival of patients with esophageal squamous cell carcinoma and gastric adenocarcinoma.
PLCE1 mRNA and protein expression and survival of patients with esophageal squamous cell carcinoma and gastric adenocarcinoma.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2014-05-29)
Wen-Qing Li, Nan Hu, Victoria H Burton, Howard H Yang, Hua Su, Catherine M Conway, Lemin Wang, Chaoyu Wang, Ti Ding, Yi Xu, Carol Giffen, Christian C Abnet, Alisa M Goldstein, Stephen M Hewitt, Philip R Taylor
ABSTRACT
Germline genetic variants in PLCE1 (10q23) have demonstrated consistent associations with risk of esophageal squamous cell carcinoma (ESCC) and gastric cancer among Chinese. We evaluated PLCE1 mRNA and protein expression in paired tumor-normal tissues, and their relationship with survival. PLCE1 mRNA was profiled using three probes in the Affymetrix GeneChip U133 for paired tumor-normal tissues of ESCC (n = 132), gastric cardia adenocarcinoma (GCA, n = 62), and gastric noncardia adenocarcinoma (GNCA, n = 72). We used immunohistochemistry to detect PLCE1 protein on slides from tissue microarrays in paired tumor-normal tissues of ESCC (n = 303), and tumors of GCA (n = 298) and GNCA (n = 124). Compared with normal tissues, PLCE1 mRNA expression was significantly reduced in ESCC tumors (P = 0.03, probe_205112_at), as well as in GCA and GNCA tumors (P < 0.0001, each probe). Protein expression was nonsignificantly reduced in ESCC tumors (P = 0.51). Increased tumor-normal mRNA fold change (probe_205112_at) was associated with longer survival in ESCC (9.6 months for highest vs. lowest quartile; Ptrend = 0.02). Increased mRNA tumor-normal fold change (probe_205111_at) was associated with longer survival for GCA (10.7 months for highest quartile; Ptrend = 0.04), but not for GNCA cases (P = 0.72). Similar to mRNA, elevated tumor-normal fold change for protein in ESCC was also associated with improved survival (8.1 months for highest quartile; Ptrend = 0.04). Dysregulated PLCE1 mRNA expression was observed for both ESCC (one probe only) and GCA tumors, and the altered PLCE1 expression seems to be associated with cancer prognosis. A potential role for PLCE1 in the early detection and/or therapy of ESCC and GCA warrants further investigation.
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material