  • Search Results
  • The cytoprotective effects of endogenous activated protein C reduce activation of coagulation during murine pneumococcal pneumonia and sepsis.
The cytoprotective effects of endogenous activated protein C reduce activation of coagulation during murine pneumococcal pneumonia and sepsis.
Thrombosis research (2015-01-15)
Marcel Schouten, Cornelis van 't Veer, Nadia Poulussen, Joost C M Meijers, Marcel Levi, Charles T Esmon, Tom van der Poll
ABSTRACT
Streptococcus (S.) pneumoniae is the most common causative pathogen in community-acquired pneumonia and sepsis. Activated protein C (APC) has been implicated as an important anticoagulant and anti-inflammatory mediator. We here sought to determine the role of the anticoagulant and cytoprotective functions of endogenous APC during pneumonia and sepsis caused by S. pneumoniae. Mice were treated intraperitoneally with monoclonal antibody (mAb) 1609 (which inhibits both anticoagulant and cytoprotective effects of APC), mAb 1591 (which inhibits only the anticoagulant effects of APC) or a control antibody mAb prior to infection with viable S. pneumoniae via the airways (to induce pneumonia) or via the tail vein (to induce primary sepsis). Mice were analyzed at 24 or 48 hours after infection. mAb 1609, but not mAb 1591, enhanced the procoagulant response to pneumococcal pneumonia and sepsis, as indicated by elevated levels of thrombin-antithrombin complexes and D-dimer in plasma and lungs. mAb 1609 only modestly affected the fibrinolytic response (elevated plasma and lung levels of the fibrinolysis inhibitor plasminogen activator inhibitor type I during sepsis) and cytokine release (elevated plasma interleukin-6 concentrations during pneumonia). The cytoprotective effects of endogenous APC reduce activation of coagulation during murine pneumococcal pneumonia and sepsis.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Pefabloc® SC, analytical standard
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis