  • Search Results
  • Development of a high-throughput assay for human proprotein convertase 5/6 for detecting uterine receptivity.
Development of a high-throughput assay for human proprotein convertase 5/6 for detecting uterine receptivity.
Analytical biochemistry (2015-01-03)
Sophea Heng, Kemperly Dynon, Ying Li, Tracey Edgell, Kelly Walton, Luk J Rombauts, Beverley Vollenhoven, Guiying Nie
ABSTRACT
Embryo implantation requires a healthy embryo and a receptive uterus. In women, the inner lining of the uterus, the endometrium, remains in a hostile state and becomes receptive for embryo implantation for only a short period during each menstrual cycle. Determining endometrial receptivity is vital in in vitro fertilization (IVF) treatment because the timing of embryo transfer needs to be synchronized with endometrial receptivity. We have previously demonstrated that proprotein convertase 5/6A (PC6) is highly expressed in the receptive endometrium and that PC6 is critical for receptivity establishment in women. Furthermore, endometrial PC6 is secreted into the uterine fluid, and levels correlate with receptivity status. Detection of PC6 in uterine fluids, therefore, would provide a nonsurgical assessment of endometrial receptivity. However, to date no assays are available for human PC6. In this study, we produced three PC6 monoclonal antibodies (mAbs) and developed a sandwich enzyme-linked immunosorbent assay (ELISA) for PC6 detection in human uterine fluids. The PC6 mAbs were confirmed to be highly specific to PC6, and the ELISA detected PC6 in human uterine fluids with a significantly higher level during the receptive phase. This newly established PC6 ELISA provides an important tool in the development of noninvasive strategies to detect endometrial receptivity in women.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Supelco
Hematoxylin
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.