  • Search Results
  • Exploring the Group B Streptococcus capsular polysaccharides: the structural diversity provides the basis for development of NMR-based identity assays.
Exploring the Group B Streptococcus capsular polysaccharides: the structural diversity provides the basis for development of NMR-based identity assays.
Journal of pharmaceutical and biomedical analysis (2014-05-31)
Vittoria Pinto, Francesco Berti
ABSTRACT
Carbohydrate-based vaccines constitute a potent tool for prevention of life-threatening bacterial infectious diseases like meningitis and pneumonia. Group B Streptococcus (GBS) is a major cause of neonatal sepsis and meningitis, particularly in infants born from mothers carrying the bacteria, and no vaccine is currently available. High-field Nuclear Magnetic Resonance (NMR) spectroscopy has been found to be an extremely robust tool for tracking the industrial process manufacturing of carbohydrate-based vaccines. Here we review the differences in the repeating unit structures of GBS capsular polysaccharide (CPS) type (Ia, Ib, II-VIII) yielding unique NMR proton profiles. All the profiles provided opportunities for selecting well resolved signals, in particular in the anomeric, the methylene protons at position C3 of N-acetyl-neuraminic acid (NeupNAc) and the N-Acetyl regions, which could be employed to develop an identity assay for monovalent vaccine bulks. Finally we reported a preliminary proof of concept of identity testing on a GBS CPS type Ia, Ib, III trivalent vaccine as blended bulks, based on the selection of one specific signal for each type in the anomeric region.
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D