  • Search Results
  • Stereoselective synthesis of the C13-C22 fragment of callystatin A by a non-aldol approach.
Stereoselective synthesis of the C13-C22 fragment of callystatin A by a non-aldol approach.
Organic & biomolecular chemistry (2015-04-02)
Sadagopan Raghavan, Sheelamanthula Rajendar
ABSTRACT
An efficient synthetic route to the C13-C22 subunit of callystatin A is reported. The key features include diastereoselective alkylation, using Myers auxiliary, for the preparation of the three carbon synthon 7, stereo- and regioselective oxidative vicinal functionalization of an electron deficient trisubstituted (Z)-olefin using an intramolecular sulfinyl group as the nucleophile, diastereoselective radical debromination of a bromohydrin derivative using Guindon's protocol to prepare the C16-C18 anti-anti stereotriad, Lewis acid promoted crotylation following Keck's protocol to create C19, C20 stereocenters and the use of the Pummerer reaction to reveal an aldehyde for the extension of two carbons by Wittig olefination.
Product Description
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Potassium permanganate, puriss. p.a., ACS reagent, Hg ≤0.000005%, ≥99.0% (RT)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Potassium permanganate, ≤150 μm particle size, 97%
Sigma-Aldrich
Potassium permanganate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%