  • Search Results
  • Natural killer cells play a critical role in cardiac allograft vasculopathy in an interleukin-6--dependent manner.
Natural killer cells play a critical role in cardiac allograft vasculopathy in an interleukin-6--dependent manner.
Transplantation (2014-10-07)
Zhu-Xu Zhang, Xuyan Huang, Jifu Jiang, Dameng Lian, Wei-Ping Min, Weihua Liu, Aaron Haig, Anthony M Jevnikar
ABSTRACT
Approximately 50% of cardiac transplants fail in the long term, and currently, there are no specific treatments to prevent chronic rejection. In the clinic, donor cardiac graft ischemia time is limited to within a few hours and correlates with delayed graft function and organ failure. It is still unknown how ischemic injury negatively influences allograft function over the long term despite advances in immunosuppression therapy. Allogeneic cardiac grafts were stored at 4 °C for 4 hr before being transplanted into T/B cell-deficient Rag(-/-) mice or T/B/natural killer (NK) cell-deficient γc(-/-)Rag(-/-) mice. Grafts were harvested 60 days after transplantation and indicators of chronic allograft vasculopathy (CAV) were quantified. We have found that cold ischemia of cardiac grafts induces CAV after transplantation into Rag1(-/-) mice. Interestingly, cold ischemia-induced CAV posttransplantation was not seen in T/B/NK cell-deficient γc(-/-)Rag(-/-) mice. However, cardiac grafts in γc(-/-)Rag(-/-) mice that received an adoptive transfer of NK cells developed CAV, supporting the role of NK cells in CAV development. Analysis of various cytokines that contribute to NK cell function revealed high interleukin (IL)-6 expression in cardiac grafts with CAV. In addition, IL-6-deficient cardiac grafts did not develop CAV after transplantation into allogeneic Rag(-/-) mice. These data demonstrate that cold ischemia and NK cells play critical roles in the development CAV. Natural killer cells and injured grafts may play a reciprocal role for CAV development in an IL-6-independent manner. Specific therapeutic strategies may be required to attenuate NK cell contribution to chronic cardiac rejection.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
L-Ascorbic acid, analytical standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture