  • Search Results
  • The ap-2 clathrin adaptor mediates endocytosis of an inhibitory killer cell Ig-like receptor in human NK cells.
The ap-2 clathrin adaptor mediates endocytosis of an inhibitory killer cell Ig-like receptor in human NK cells.
Journal of immunology (Baltimore, Md. : 1950) (2014-09-23)
Amanda K Purdy, Diana A Alvarez Arias, Jennifer Oshinsky, Ashley M James, Ilya Serebriiskii, Kerry S Campbell
ABSTRACT
Stable surface expression of human inhibitory killer cell Ig-like receptors (KIRs) is critical for controlling NK cell function and maintaining NK cell tolerance toward normal MHC class I(+) cells. Our recent experiments, however, have found that Ab-bound KIR3DL1 (3DL1) readily leaves the cell surface and undergoes endocytosis to early/recycling endosomes and subsequently to late endosomes. We found that 3DL1 internalization is at least partially mediated by an interaction between the μ2 subunit of the AP-2 clathrin adaptor complex and ITIM tyrosine residues in the cytoplasmic domain of 3DL1. Disruption of the 3DL1/μ2 interaction, either by mutation of the ITIM tyrosines in 3DL1 or mutation of μ2, significantly diminished endocytosis and increased surface expression of 3DL1 in human primary NK cells and cell lines. Furthermore, we found that the 3DL1/AP-2 interaction is diminished upon Ab engagement with the receptor, as compared with untreated cells. Thus, we have identified AP-2-mediated endocytosis as a mechanism regulating the surface levels of inhibitory KIRs through their ITIM domains. Based on our results, we propose a model in which nonengaged KIRs are internalized by this mechanism, whereas engagement with MHC class I ligand would diminish AP-2 binding, thereby prolonging stable receptor surface expression and promoting inhibitory function. Furthermore, this ITIM-mediated mechanism may similarly regulate the surface expression of other inhibitory immune receptors.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
IL-2 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)