  • Search Results
  • Normalization of urinary pteridines by urine specific gravity for early cancer detection.
Normalization of urinary pteridines by urine specific gravity for early cancer detection.
Clinica chimica acta; international journal of clinical chemistry (2014-05-06)
Casey Burton, Honglan Shi, Yinfa Ma
ABSTRACT
Urinary biomarkers, such as pteridines, require normalization with respect to an individual's hydration status and time since last urination. Conventional creatinine-based corrections are affected by a multitude of patient factors whereas urine specific gravity (USG) is a bulk specimen property that may better resist those same factors. We examined the performance of traditional creatinine adjustments relative to USG to six urinary pteridines in aggressive and benign breast cancers. 6-Biopterin, neopterin, pterin, 6-hydroxymethylpterin, isoxanthopterin, xanthopterin, and creatinine were analyzed in 50 urine specimens with a previously developed liquid chromatography-tandem mass spectrometry technique. Creatinine and USG performance were evaluated with non-parametric Mann-Whitney hypothesis testing. USG and creatinine were moderately correlated (r=0.857) with deviations occurring in dilute and concentrated specimens. In 48 aggressive and benign breast cancers, normalization by USG significantly outperformed creatinine adjustments which marginally outperformed uncorrected pteridines in predicting pathological status. In addition, isoxanthopterin and xanthopterin were significantly higher in pathological specimens when normalized by USG. USG, as a bulk property, can provide better performance over creatinine-based normalizations for urinary pteridines in cancer detection applications.
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Pterine, ~95%
Sigma-Aldrich
6-Biopterin, ≥97%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Supelco
Potassium Iodide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis