  • Search Results
  • Preparation, characterization and catalytic activity of NiOx and NiOx/ZrO2 for oxidation of phenol in aqueous solution.
Preparation, characterization and catalytic activity of NiOx and NiOx/ZrO2 for oxidation of phenol in aqueous solution.
Acta chimica Slovenica (2015-01-01)
Dimitar Petrov, Stoyanka Christoskova, Maria Stoyanova, Vanina Ivanova, Daniela Karashanova
ABSTRACT
In the present study bulk NiO(x) and supported NiO(x)/ZrO(2) catalysts have been prepared. The bulk NiO(x) was synthesized using the precipitation-oxidation method with reverse order of precipitation while deposition-precipitation technique was used for the preparation of zirconia supported catalyst. The as-prepared samples were characterized by means of XRD, HRTEM, SAED, IR-spectroscopy, and chemical analyses. It was found that under the applied synthesis procedure nanosized and highly dispersed oxide materials with high active oxygen content were obtained. The catalytic activity and selectivity of these oxide catalysts have been studied in reaction of low-temperature oxidation of phenol in aqueous phase. The effects of several parameters such as catalyst amount, temperature and solution pH on the degradation efficiency of the process were also investigated. Experimental results demonstrated that phenol could be completely degraded under all reaction conditions, except at pH = 12.
Sigma-Aldrich
Potassium bromide, FT-IR grade, ≥99% trace metals basis
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, ≥99%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Phenol, natural, 97%, FG
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)