  • Search Results
  • A labdane diterpene exerts ex vivo and in vivo cardioprotection against post-ischemic injury: involvement of AKT-dependent mechanisms.
A labdane diterpene exerts ex vivo and in vivo cardioprotection against post-ischemic injury: involvement of AKT-dependent mechanisms.
Biochemical pharmacology (2015-01-06)
Irene Cuadrado-Berrocal, María V Gómez-Gaviro, Yolanda Benito, Alicia Barrio, Javier Bermejo, María Eugenia Fernández-Santos, Pedro L Sánchez, Manuel Desco, Francisco Fernández-Avilés, María Fernández-Velasco, Lisardo Boscá, Beatriz de Las Heras
ABSTRACT
Therapeutic approaches to protect the heart from ischemia/reperfusion (I/R) injury are an area of intense research, as myocardial infarction is a major cause of mortality and morbidity. Diterpenes are bioactive natural products with great therapeutic potential. In the present study, we have investigated the in vivo cardioprotective effects of a labdane diterpene (DT1) against cardiac I/R injury and the molecular mechanisms involved. DT1 attenuates post-ischemic injury via an AKT-dependent activation of HIF-1α, survival pathways and inhibition of NF-κB signaling. Myocardial infarction (MI) was induced in Wistar rats occluding the left coronary artery (LCA) for 30min followed by 72h reperfusion. DT1 (5mg/kg) was intravenously administered at reperfusion. In addition, we investigated the mechanisms of cardioprotection in the Langendorff-perfused model. Cardioprotection was observed when DT1 was administered after myocardial injury. The molecular mechanisms involved the activation of the survival pathway PDK-1, AKT and AMPK, a reduced phosphorylation of PKD1/2 and sustained HIF-1α activity, leading to increased expression of anti-apoptotic proteins and decreased caspase-3 activation. Pharmacological inhibition of AKT following MI and prior to DT1 challenge significantly decreased the cardioprotection afforded by DT1 therapy at reperfusion. Cardiac function after MI was significantly improved after DT1-treatment, as evidenced by hemodynamic recovery and decreased myocardial infarct size. These findings demonstrate an efficient in vivo cardioprotection by diterpene DT1 against I/R when administered at reperfusion, opening new therapeutic strategies as adjunctive therapy for the pharmacological management of I/R injury.
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sevoflurane, European Pharmacopoeia (EP) Reference Standard