  • Search Results
  • Electrogenic and nonelectrogenic ion fluxes across lipid and mitochondrial membranes mediated by monensin and monensin ethyl ester.
Electrogenic and nonelectrogenic ion fluxes across lipid and mitochondrial membranes mediated by monensin and monensin ethyl ester.
Biochimica et biophysica acta (2015-01-21)
Yuri N Antonenko, Tatyana I Rokitskaya, Adam Huczyński
ABSTRACT
Monensin is a carrier of cations through lipid membranes capable of exchanging sodium (potassium) cations for protons by an electroneutral mechanism, whereas its ethyl ester derivative ethyl-monensin is supposed to transport sodium (potassium) cations in an electrogenic manner. To elucidate mechanistic details of the ionophoric activity, ion fluxes mediated by monensin and ethyl-monensin were measured on planar bilayer lipid membranes, liposomes, and mitochondria. In particular, generation of membrane potential on liposomes was studied via the measurements of rhodamine 6G uptake by fluorescence correlation spectroscopy. In mitochondria, swelling experiments were expounded by the additional measurements of respiration, membrane potential, and matrix pH. It can be concluded that both monensin and ethyl-monensin can perform nonelectrogenic exchange of potassium (sodium) ions for protons and serve as electrogenic potassium ion carriers similar to valinomycin. The results obtained are in line with the predictions based on the crystal structures of the monensin complexes with sodium ions and protons (Huczyński et al., Biochim. Biophys. Acta, 1818 (2012) pp. 2108-2119). The functional activity observed for artificial membranes and mitochondria can be applied to explain the activity of ionophores in living systems. It can also be important for studying the antitumor activity of monensin.
Sigma-Aldrich
8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt, ≥96%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium hydride, in paraffin
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis