  • Search Results
  • TLR2/TLR4-dependent exaggerated cytokine production in hyperimmunoglobulinaemia D and periodic fever syndrome.
TLR2/TLR4-dependent exaggerated cytokine production in hyperimmunoglobulinaemia D and periodic fever syndrome.
Rheumatology (Oxford, England) (2014-09-01)
Monique Stoffels, Johanna Jongekrijg, Thijs Remijn, Nina Kok, Jos W M van der Meer, Anna Simon
ABSTRACT
The autoinflammatory hyperimmunoglobulinaemia D and periodic fever syndrome (HIDS) is characterized by recurrent episodes of fever and inflammation. As part of the mevalonate kinase deficiency spectrum, it is caused by MVK mutations, resulting in decreased mevalonate kinase activity in the isoprenoid pathway. Although IL-1β is considered a major cytokine in its pathogenesis, IL-1 blockade is not successful in a proportion of patients. We aimed to further characterize the pro-inflammatory cytokine profile of HIDS. Peripheral blood mononuclear cells from HIDS patients and healthy donors were incubated with several stimuli. Cytokine concentrations were detected by ELISA. To analyse mRNA and protein expression, we performed quantitative RT-PCR and western blot, respectively. We observed significant differences in cytokine production when cells were incubated with ligands for Toll-like receptor 2 (TLR2), TLR4 and nucleotide-binding oligomerization domain-containing 2 (NOD2). The increased ratio between active and inactive caspase-1 protein in HIDS patients could explain why these cells are more easily triggered to secrete IL-1β. This is apparently not regulated at the transcriptional level, since expression levels of caspase-1 and IL-1β mRNA were similar in patients and controls. Both anakinra and tocilizumab treatment resulted in decreased inflammation, both ex vivo as well as in vivo. The increased cytokine secretion in HIDS is specific for TLR2, TLR4 and NOD2 ligation. Although IL-1β is important in the HIDS pathology, our data suggest it is a multicytokine disease. A more rigorous clinical trial is required to determine whether IL-6 receptor blockade may be considered in patients not responding to anakinra treatment.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%