  • Search Results
  • Antimicrobial and enzyme inhibitory activities of the constituents of Plectranthus madagascariensis (Pers.) Benth.
Antimicrobial and enzyme inhibitory activities of the constituents of Plectranthus madagascariensis (Pers.) Benth.
Journal of enzyme inhibition and medicinal chemistry (2014-02-11)
Renata Kubínová, Radka Pořízková, Alice Navrátilová, Oldřich Farsa, Zuzana Hanáková, Adriana Bačinská, Alois Cížek, Marie Valentová
ABSTRACT
Plectranthus madagascariensis is used as a traditional medicine in Southern Africa. In search of compounds and activities supporting the medicinal use, the chemical profile of the methanolic extract was studied by high-performance liquid chromatography with diode array detection (HPLC-DAD). Four major constituents were isolated and identified as rosmarinic acid (1), 7β,6β-dihydroxyroyleanone (2), 7β-acetoxy-6β-hydroxyroyleanone (3) and coleon U quinone (4). The two abietane diterpenoids (2 and 3) were isolated for the first time from this species. Antimicrobial, cholinesterase and α-glucosidase inhibitory activities of these compounds were studied. The compounds exhibited inhibitory activity on α-glucosidase with IC50 values from 33 to 275 μM. Abietanes showed potent antibacterial activity against Staphylococcus aureus and Enterococcus faecalis.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
Acarbose, ≥95%
Sigma-Aldrich
Rosmarinic acid, 96%
Supelco
Acarbose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Sigma-Aldrich
Butyrylthiocholine iodide, ≥98%
Sigma-Aldrich
Butyrylthiocholine iodide, puriss., ≥99.0% (AT)
Rosmarinic acid, European Pharmacopoeia (EP) Reference Standard
Rosmarinic acid, primary reference standard
Acarbose, European Pharmacopoeia (EP) Reference Standard