  • Search Results
  • Characterization of the enzymatic and structural properties of human D-aspartate oxidase and comparison with those of the rat and mouse enzymes.
Characterization of the enzymatic and structural properties of human D-aspartate oxidase and comparison with those of the rat and mouse enzymes.
Biological & pharmaceutical bulletin (2015-03-10)
Masumi Katane, Tomonori Kawata, Kazuki Nakayama, Yuki Saitoh, Yuusuke Kaneko, Satsuki Matsuda, Yasuaki Saitoh, Tetsuya Miyamoto, Masae Sekine, Hiroshi Homma
ABSTRACT
D-Aspartate (D-Asp), a free D-amino acid found in mammals, plays crucial roles in the neuroendocrine, endocrine, and central nervous systems. Recent studies have implicated D-Asp in the pathophysiology of infertility and N-methyl-D-Asp receptor-related diseases. D-Asp oxidase (DDO), a degradative enzyme that is stereospecific for acidic D-amino acids, is the sole catabolic enzyme acting on D-Asp in mammals. Human DDO is considered an attractive therapeutic target, and DDO inhibitors may be potential lead compounds for the development of new drugs against the aforementioned diseases. However, human DDO has not been characterized in detail and, although preclinical studies using experimental rodents are prerequisites for evaluating the in vivo effects of potential inhibitors, the existence of species-specific differences in the properties of human and rodent DDOs is still unclear. Here, the enzymatic activity and characteristics of purified recombinant human DDO were analyzed in detail. The kinetic and inhibitor-binding properties of this enzyme were also compared with those of purified recombinant rat and mouse DDOs. In addition, structural models of human, rat, and mouse DDOs were generated and compared. It was found that the differences among these DDO proteins occur in regions that appear involved in migration of the substrate/product in and out of the active site. In summary, detailed characterization of human DDO was performed and provides useful insights into the use of rats and mice as experimental models for evaluating the in vivo effects of DDO inhibitors.
Product Description
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M