  • Search Results
  • Sex- and smoke-related differences in gastrointestinal transit of cyclosporin A microemulsion capsules.
Sex- and smoke-related differences in gastrointestinal transit of cyclosporin A microemulsion capsules.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2014-07-23)
Pietro Fagiolino, Marta Vázquez, Manuel Ibarra, Laura Magallanes, Natalia Guevara, Nikoletta Fotaki
ABSTRACT
The aim of this work was to study the effect of the sex and the smoking status on the pharmacokinetics and the bioequivalence assessment of a branded and a generic cyclosporine A microemulsion formulation in soft-gelatin capsule. Sixteen healthy volunteers (eight women and eight men) participated in a CyA bioequivalence study, with nine of the volunteers being smokers. Sandimmun Neoral® (brand formulation; Reference) and Sigmasporin Microral® (generic formulation; Test) were administered under fasting conditions. Pharmacokinetic parameters were calculated through non compartmental analysis. Bioequivalence was declared based on the 90% confidence intervals (90% CI) for the T/R ratio of the geometric means for each parameter. In vitro determination of the capsules opening time was performed in simulated gastric fluid without enzyme with USP Apparatus 2. The extent of absorption was similar between both products for all subjects or each sex-group. The absorption rate was similar for both products when considering all subjects, whereas a significant difference in the TMAX between the two products was observed for the male subjects only, which relates to its slower capsule opening time observed in vitro (12.4 versus 6.0 min). No differences were observed in women that could relate to their slower gastric emptying. Differences in drug exposure were observed between smokers and non-smokers. Sex- and smoke-related differences in the gastrointestinal transit should be considered when the on-set time would be determinant for the treatment success of a drug.
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al