  • Search Results
  • Determination of receptor protein binding site specificity and relative binding strength using a time-resolved competition assay.
Determination of receptor protein binding site specificity and relative binding strength using a time-resolved competition assay.
Journal of pharmacological and toxicological methods (2014-08-02)
Pavel Barta, Marie Volkova, Adrian Dascalu, Diana Spiegelberg, Frantisek Trejtnar, Karl Andersson
ABSTRACT
Competitive binding assays can be used to decipher not only the binding kinetics of studied ligands but also the binding site preference. Such assays are an essential step in the characterization of radioligands. However, the currently used competition assays require high concentrations of usually expensive ligands and still provide only binding site preference. By employing the time-resolved competition assay presented in this paper, binding characteristics including binding site preference can be obtained using less ligand. To demonstrate the appropriateness of the time-resolved competition assay, we developed an assay in which the ligand binding was interrupted with a competitor. Experiments were performed on human carcinoma cell lines expressing epidermal growth factor receptor (EGFR). The targeting of the receptor was performed with radio-iodinated epidermal growth factor (EGF). The employed competitors involved either natural ligand transforming growth factor alpha (TGF-α) or anti-EGFR antibodies cetuximab and panitumumab targeting the same EGFR domain. Radio-iodinated EGF bound to EGFR was displaced with either low concentrations of cetuximab or high concentrations of panitumumab. In the case of TGF-α, we observed no competitive displacement of bound EGF at either high or low concentrations. When comparing the time-resolved competition assay with a manual competition assay, the resulting data of measured inhibition constants were in agreement. The results summarised in this study confirm the appropriateness of the time-resolved competition assay for assessing ligand binding properties. The assay has the potential to complement or replace conventional competition assays for determining binding site preference in the future.
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Anti-EGFR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 102618, purified immunoglobulin, lyophilized powder
Sigma-Aldrich
Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 29.1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone F4, ascites fluid
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Monoclonal Anti-EGFR antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-EGFR antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0815, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Rabbit anti-EGFR Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets