  • Search Results
  • Relationships between T cell and IgE/IgG4 epitopes of the Anisakis simplex major allergen Ani s 1.
Relationships between T cell and IgE/IgG4 epitopes of the Anisakis simplex major allergen Ani s 1.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology (2014-12-17)
M Garcia Alonso, M L Caballero, A Umpierrez, M Lluch-Bernal, T Knaute, R Rodríguez-Pérez
ABSTRACT
Anisakiasis is a global disease caused by the consumption of raw or lightly cooked fish parasitized with third-stage Anisakis larvae. Anisakis simplex allergens may cause severe allergic reactions including angio-oedema, urticaria and anaphylaxis. Approximately 80% of allergic patients have allergen-specific IgE against Ani s 1, and the diagnostic value of testing for antibodies to Ani s 1 has been extensively demonstrated. However, no previous studies have investigated the molecular aspects of the allergic response to Ani s 1. Knowledge of allergen-specific T cell and B cell (IgE and IgG4) epitopes is important for elucidating the immunological mechanisms underlying allergic responses, and for understanding why particular proteins behave as allergens. To elucidate the main T cell- and B cell (IgE and IgG4)- binding regions of Ani s 1. T cell epitopes were identified by peptide proliferation assays using T cell lines derived from peripheral blood mononuclear cells of 11 patients with Anisakis allergy, and IgE and IgG4 epitopes were identified by microarray immunoassay using sera from a different group of 11 patients with Anisakis allergy. Several T cell epitopes of Ani s 1 were identified, of which Ani s 1145-156 , Ani s 1151-162 and Ani s 1163-171 located at the C-terminal end of the protein were the most relevant. IgE and IgG4 recognized largely the same peptides, including Ani s 122-41 , Ani s 125-44 , Ani s 127-47 , Ani s 137-56 and Ani s 194-113 . This is the first report describing the T cell epitopes of an important allergen of A. simplex, and the first B cell epitope study of this allergen in the Spanish population. This information can help to elucidate the mechanisms underlying the allergic response to Ani s 1, potentially leading to therapeutic and diagnostic advances.
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Sigma-Aldrich
IL-2 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)