  • Search Results
  • Inhibition of protein kinase C enhances angiogenesis induced by platelet-derived growth factor C in hyperglycemic endothelial cells.
Inhibition of protein kinase C enhances angiogenesis induced by platelet-derived growth factor C in hyperglycemic endothelial cells.
Cardiovascular diabetology (2015-04-08)
Junji Moriya, Napoleone Ferrara
ABSTRACT
Diabetes is a risk factor for the development of cardiovascular diseases with impaired angiogenesis. We have previously shown that platelet-derived growth factor C (PDGF-C) and its receptor, PDGF receptor α (PDGFR-α) were downregulated in ischemic limbs of diabetic mice, although the underlying mechanisms remained elusive. Protein kinase C (PKC) is a family of serine/threonine kinases and is known to be involved in angiogenesis. The purpose of this study is to elucidate the mechanisms of how PDGF-C/PDGFR-α axis is impaired in diabetes. Human umbilical vein endothelial cells (HUVECs) and human cardiac microvascular endothelial cells (HMVECs) cultured in normoglycemic or hyperglycemic conditions were examined. We also examined the effects of PKC inhibition on the PDGF-C/PDGFR-α axis in endothelial cells exposed to hyperglycemia. Hyperglycemia inhibited proliferation and decreased viability of both HUVECs and HMVECs. Hyperglycemic endothelial cells exhibited decreased PDGFR-α expression both at messenger RNA (mRNA) and protein levels, while there was no significant change in expression of PDGF-C. We also found that expression of PKC-α, one of the PKC isoforms, was increased in hyperglycemic endothelial cells and that inhibition of PKC upregulated PDGFR-α expression in these cells. Phosphorylation of extracellular signal-regulated kinase (ERK) and Akt induced by PDGF-C was significantly attenuated in hyperglycemic endothelial cells, whereas inhibition of PKC effectively reversed these inhibitory effects. Moreover, inhibition of PKC also promoted angiogenesis induced by PDGF-C in hyperglycemic endothelial cells, which was not observed in vascular endothelial growth factor-A (VEGF-A)-induced angiogenesis. These findings suggest that downregulation of the PDGF-C/PDGFR-α axis is involved in impaired angiogenesis of hyperglycemia through upregulation of PKC. Targeting PKC to restore PDGF-C signaling might be a novel therapeutic strategy for the treatment of vascular complications in diabetes.
Product Description
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
DL-Sulforaphane, ≥90% (HPLC), synthetic, liquid
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
GF 109203X, synthetic, ≥90% (HPLC)
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)