  • Search Results
  • Octarepeat region flexibility impacts prion function, endoproteolysis and disease manifestation.
Octarepeat region flexibility impacts prion function, endoproteolysis and disease manifestation.
EMBO molecular medicine (2015-02-11)
Agnes Lau, Alex McDonald, Nathalie Daude, Charles E Mays, Eric D Walter, Robin Aglietti, Robert C C Mercer, Serene Wohlgemuth, Jacques van der Merwe, Jing Yang, Hristina Gapeshina, Chae Kim, Jennifer Grams, Beipei Shi, Holger Wille, Aru Balachandran, Gerold Schmitt-Ulms, Jiri G Safar, Glenn L Millhauser, David Westaway
ABSTRACT
The cellular prion protein (PrP(C)) comprises a natively unstructured N-terminal domain, including a metal-binding octarepeat region (OR) and a linker, followed by a C-terminal domain that misfolds to form PrP(S) (c) in Creutzfeldt-Jakob disease. PrP(C) β-endoproteolysis to the C2 fragment allows PrP(S) (c) formation, while α-endoproteolysis blocks production. To examine the OR, we used structure-directed design to make novel alleles, 'S1' and 'S3', locking this region in extended or compact conformations, respectively. S1 and S3 PrP resembled WT PrP in supporting peripheral nerve myelination. Prion-infected S1 and S3 transgenic mice both accumulated similar low levels of PrP(S) (c) and infectious prion particles, but differed in their clinical presentation. Unexpectedly, S3 PrP overproduced C2 fragment in the brain by a mechanism distinct from metal-catalysed hydrolysis reported previously. OR flexibility is concluded to impact diverse biological endpoints; it is a salient variable in infectious disease paradigms and modulates how the levels of PrP(S) (c) and infectivity can either uncouple or engage to drive the onset of clinical disease.
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Guanidine hydrochloride solution
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Proline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, ≥99% (titration), organic base and chaeotropic agent
Sigma-Aldrich
DL-Phenylalanine, ReagentPlus®, 99%
Sigma-Aldrich
DL-Proline, ReagentPlus®, 99%
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Phenylalanine, European Pharmacopoeia (EP) Reference Standard
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
SAFC
Guanidine hydrochloride
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
Sigma-Aldrich
1,1′-Carbonyldiimidazole solution, 0.4 M in methylene chloride