  • Search Results
  • Evidence that intraspecific trait variation among nasal bacteria shapes the distribution of Staphylococcus aureus.
Evidence that intraspecific trait variation among nasal bacteria shapes the distribution of Staphylococcus aureus.
Infection and immunity (2014-07-02)
Ben Libberton, Rosanna E Coates, Michael A Brockhurst, Malcolm J Horsburgh
ABSTRACT
Nasal carriage of Staphylococcus aureus is a risk factor for infection, yet the bacterial determinants required for carriage are poorly defined. Interactions between S. aureus and other members of the bacterial flora may determine colonization and have been inferred in previous studies by using correlated species distributions. However, traits mediating species interactions are often polymorphic, suggesting that understanding how interactions structure communities requires a trait-based approach. We characterized S. aureus growth inhibition by the culturable bacterial aerobe consortia of 60 nasal microbiomes, and this revealed intraspecific variation in growth inhibition and that inhibitory isolates clustered within communities that were culture negative for S. aureus. Across microbiomes, the cumulative community-level growth inhibition was negatively associated with S. aureus incidence. To fully understand the ecological processes structuring microbiomes, it will be crucial to account for intraspecific variation in the traits that mediate species interactions.
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
D-Mannitol, ACS reagent
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Mannitol, European Pharmacopoeia (EP) Reference Standard