  • Search Results
  • Doxycycline exerts multiple anti-allergy effects to attenuate murine allergic conjunctivitis and systemic anaphylaxis.
Doxycycline exerts multiple anti-allergy effects to attenuate murine allergic conjunctivitis and systemic anaphylaxis.
Biochemical pharmacology (2014-08-17)
Wenru Su, Qian Wan, Longhui Han, Jingwen Huang, Xiaoqing Chen, Guihua Chen, Song Guo Zheng, Dan Liang
ABSTRACT
Allergic diseases, which affect up to 20-30% of the world population, are still therapeutic challenge for allergists. Tetracyclines, which belong to an antibiotic drug family that possesses a striking variety of non-antibiotic properties, have been successfully applied to a wide range of diseases. However, their roles in allergic conjunctivitis and anaphylaxis and their underlying anti-allergy mechanisms remain elusive. Here, we reported that treatment with doxycycline significantly reduced IgE release from mouse B cells and the degranulation and inflammatory cytokines production of mouse mast cells (MCs) activated by IgE-dependent way. Furthermore, doxycycline treatment significantly inhibited histamine-induced vascular hyperpermeability in vitro. Mechanistically, the doxycycline-mediated inhibition of B cells, MCs and histamine may occur via modulation of the PI3K/Akt pathway. In vivo, our results demonstrated that treatment with doxycycline significantly attenuated clinical symptoms of mouse models of experimental allergic conjunctivitis (EAC) with a significant decrease in inflammatory cell frequency, IgE production, histamine release, and a decrease in TNF-α and IL-4 production. Using mouse models of MCs-dependent passive systemic anaphylaxis (PSA), we further confirmed anti-allergy effects of doxycycline and doxycycline-mediated inhibitory effects on MCs. Furthermore, our results showed that doxycycline significantly attenuate histamine-induced systemic anaphylaxis-like reaction (HISA) with a significantly downregulation of PI3K/Akt/eNOS/VE-cadherin pathway. The doxycycline-mediated anti-allergy effects during EAC, PSA and HISA were abrogated when an Akt activator, SC79, was administered. These findings suggest that doxycycline inhibits B cell, MC and histamine function and attenuates experimental allergic conjunctivitis and systemic anaphylaxis by possible modulating the PI3K/Akt pathway.
Sigma-Aldrich
Monoclonal Anti-Rac antibody produced in mouse, clone 23A8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Toluidine Blue, for microscopy (Hist., Vit.)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
IL-3 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
IL-3 from mouse, Carrier free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), suitable for cell culture
Sigma-Aldrich
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
IL-3 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Anti-AKT1 (Ab-473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Rabbit anti-AKT1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Monoclonal Anti-AKT1 antibody produced in mouse, clone 2E11, purified immunoglobulin, buffered aqueous solution