A potent anticancer agent of shikonin derivative targeting tubulin.
Chirality (2015-02-11)
Shahla Karim Baloch, Lin Ma, Guo-Hua Xu, Li-Fei Bai, Hua Zhao, Cheng-Yi Tang, Yan-Jun Pang, Rong-Wu Yang, Xiao-Ming Wang, Gui-Hua Lu, Yong-Hua Yang
ABSTRACT
In this study, a shikonin ester derivative, compound , was selected to evaluate its anticancer activities and we found that compound exhibited better antitubulin activities against the human HepG2 cell line with an IC50 value of 1.097 μM. Furthermore, the inhibition of tubulin polymerization results indicated that compound demonstrated the most potent antitubulin activity (IC50  = 13.88), which was compared with shikonin and colchicine as positive controls (IC50  = 25.28 μM and 22.56 μM), respectively. Compound was simulated to have good binding site with tubulin and arrested the cell cycle at G2/M phase, which also induces apoptosis in HepG2 cells, in which P53 and members of Bcl-2 protein family were both involved in the progress of apoptosis revealed by western blot. Confocal microscopy observations revealed compound targeted tubulin and altered its polymerization by interfering with microtubule organization. Based on these results, compound functions as a potent anticancer agent targeting tubulin.
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sucrose, meets USP testing specifications