  • Search Results
  • CXCR4 and PTEN are involved in the anti-metastatic regulation of anethole in DU145 prostate cancer cells.
CXCR4 and PTEN are involved in the anti-metastatic regulation of anethole in DU145 prostate cancer cells.
Biochemical and biophysical research communications (2014-02-15)
Yun-Hee Rhee, Phil-Sang Chung, Sung-Hoon Kim, Jin Chul Ahn
ABSTRACT
Anethole has been known to have chemopreventive activities as a suppressor of the incidence and multiplicity of both invasive and noninvasive carcinomas. The goal of this study was to understand the anti-metastatic effect of anethole through C-X-C chemokine receptor type 4 (CXCR4)/tumor suppressor phosphatase and tensin homologue (PTEN) axis in DU145 prostate cancer cells. Anethole reduced both of the RNA level and the protein level of CXCR4 in a dose-dependent manner without cytotoxicity. Anethole also reduced the expression of CXCR4 and prolonged the expression of PTEN in DU145 prostate cancers. The phosphorylation of AKT and phosphatidylinositol-3kinase (PI3K) were decreased with anethole. The inhibition metastatic effect of anethole was arisen from down-regulating CXCR4 and up-regulating PTEN. Morphologically, anethole significantly inhibited the invasion of DU145 cell and down-regulated the activities of matrix-metalloproteinase (MMPs) in a dose-dependent manner. However, anethole didnot decrease the phosphorylation of PI3K and AKT while PTEN was silenced. Furthermore, the CXCR4 inhibition of anethole was not caused to proteasomal or lysosomal of CXCR4. Taken together, anethole demonstrated to act as the CXCR4 antagonist and as the PTEN activator which resulted to PI3K/AKT-mediated inhibition of the metastatic prostate cancer progressions.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)