  • Search Results
  • Natural killer and dendritic cells collaborate in the immune response induced by the vaccine against uterine cervical cancer.
Natural killer and dendritic cells collaborate in the immune response induced by the vaccine against uterine cervical cancer.
European journal of immunology (2014-09-18)
Inge Langers, Virginie Renoux, Anca Reschner, Antoine Touzé, Pierre Coursaget, Jacques Boniver, Joachim Koch, Philippe Delvenne, Nathalie Jacobs
ABSTRACT
Virus-like particles (VLPs) of human papillomavirus (HPV) are used as a vaccine against HPV-induced cancer, and recently we have shown that these VLPs are able to activate natural killer (NK) cells. Since NK cells collaborate with dendritic cells (DCs) to induce an immune response against viral infections and tumors, we studied the impact of this crosstalk in the context of HPV vaccination. NK cells in the presence of HPV-VLPs enhanced DC-maturation as shown by an upregulation of CD86 and HLA-DR and an increased production of IL-12p70, but not of the immunosuppressive cytokine IL-10. This activation was bidirectional. Indeed, in the presence of HPV-VLPs, DCs further activated NK cells by inducing the upregulation of cell surface activation markers (CD69 and HLA-DR). The function of NK cells was also improved as shown by an increase in IFN-γ secretion and cytotoxic activity against an HPV(+) cell line. This crosstalk between NK cells and DCs needed CD40 interaction and IL-12p70 secretion, whereas NKG2D was not implicated. Our results provide insight into how VLPs interact with innate immune cells and how NK cells and DCs play a role in the immune response induced by this vaccine agent.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%