  • Search Results
  • Proteome-wide lysine acetylation profiling of the human pathogen Mycobacterium tuberculosis.
Proteome-wide lysine acetylation profiling of the human pathogen Mycobacterium tuberculosis.
The international journal of biochemistry & cell biology (2014-12-03)
Longxiang Xie, Xiaobo Wang, Jie Zeng, Mingliang Zhou, Xiangke Duan, Qiming Li, Zhen Zhang, Hongping Luo, Lei Pang, Wu Li, Guojian Liao, Xia Yu, Yunxu Li, Hairong Huang, Jianping Xie
ABSTRACT
N(ɛ)-Acetylation of lysine residues represents a pivotal post-translational modification used by both eukaryotes and prokaryotes to modulate diverse biological processes. Mycobacterium tuberculosis is the causative agent of tuberculosis, one of the most formidable public health threats. Many aspects of the biology of M. tuberculosis remain elusive, in particular the extent and function of N(ɛ)-lysine acetylation. With a combination of anti-acetyllysine antibody-based immunoaffinity enrichment with high-resolution mass spectrometry, we identified 1128 acetylation sites on 658 acetylated M. tuberculosis proteins. GO analysis of the acetylome showed that acetylated proteins are involved in the regulation of diverse cellular processes including metabolism and protein synthesis. Six types of acetylated peptide sequence motif were revealed from the acetylome. Twenty lysine-acetylated proteins showed homology with acetylated proteins previously identified from Escherichia coli, Salmonella enterica, Bacillus subtilis and Streptomyces roseosporus, with several acetylation sites highly conserved among four or five bacteria, suggesting that acetylated proteins are more conserved. Notably, several proteins including isocitrate lyase involved in the persistence, virulence and antibiotic resistance are acetylated, and site-directed mutagenesis of isocitrate lyase acetylation site to glutamine led to a decrease of the enzyme activity, indicating major roles of KAc in these proteins engaged cellular processes. Our data firstly provides a global survey of M. tuberculosis acetylation, and implicates extensive regulatory role of acetylation in this pathogen. This may serve as an important basis to address the roles of lysine acetylation in M. tuberculosis metabolism, persistence and virulence.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium butyrate, 98%
Supelco
Sodium butyrate, certified reference material, TraceCERT®
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.