  • Search Results
  • A novel cantharidin analog N-benzylcantharidinamide reduces the expression of MMP-9 and invasive potentials of Hep3B via inhibiting cytosolic translocation of HuR.
A novel cantharidin analog N-benzylcantharidinamide reduces the expression of MMP-9 and invasive potentials of Hep3B via inhibiting cytosolic translocation of HuR.
Biochemical and biophysical research communications (2014-04-17)
Ji-Yeon Lee, Tae-Wook Chung, Hee-Jung Choi, Chang Hyun Lee, Jae Soon Eun, Young Taek Han, Jun-Yong Choi, So-Yeon Kim, Chang-Woo Han, Han-Sol Jeong, Ki-Tae Ha
ABSTRACT
Invasion and metastasis are major causes of malignant tumor-associated mortality. The present study aimed to investigate the molecular events underlying inhibitory effect of N-benzylcantharidinamide, a novel synthetic analog of cantharidin, on matrix metalloproteinase-9 (MMP-9)-mediated invasion in highly metastatic hepatocellular carcinoma Hep3B cells. In this investigation, among six analogs of cantharidin, only N-benzylcantharidinamide has the inhibitory action on MMP-9 expression at non-toxic dose. The MMP-9 expression and invasion of Hep3B cells were significantly suppressed by treatment of N-benzylcantharidinamide in a dose-dependent manner. On the other hand, the transcriptional activity of MMP-9 promoter and nuclear levels of NF-κB and AP-1 as the main transcriptional factors inducing MMP-9 expression were not affected by it although the level of MMP-9 mRNA was reduced by treatment of N-benzylcantharidinamide. Interestingly, the stability of MMP-9 mRNA was significantly reduced by N-benzylcantharidinamide-treatment. In addition, the cytosolic translocation of human antigen R (HuR), which results in the increase of MMP-9 mRNA stability through interaction of HuR with 3'-untranslated region of MMP-9 mRNA, was suppressed by treatment of N-benzylcantharidinamide, in a dose-dependent manner. Taken together, it was demonstrated, for the first time, that N-benzylcantharidinamide suppresses MMP-9 expression by reducing HuR-mediated MMP-9 mRNA stability for the inhibition of invasive potential in highly metastatic Hep3B cells.
Sigma-Aldrich
Anti-MMP9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Matrix Metalloproteinase-9 antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-MMP9 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-RELA (Ab-311) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-529) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-536) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-435) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-254) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-276) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-RELA (Ab-505) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Norcantharidin, solid
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Anti-RELA (Ab-468) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-MMP9 antibody produced in mouse, clone 2H4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-MMP9 antibody produced in mouse, clone 5C3, ascites fluid
Sigma-Aldrich
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0538, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0542, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-MMP9 antibody produced in mouse, clone 5G3, ascites fluid
Sigma-Aldrich
Monoclonal Anti-MMP9 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0539, purified immunoglobulin, buffered aqueous glycerol solution
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cantharidin