  • Search Results
  • Comparison of nephron-protective effects of enalapril and GLP analogues (exenatide) in diabetic nephropathy.
Comparison of nephron-protective effects of enalapril and GLP analogues (exenatide) in diabetic nephropathy.
Experimental and clinical endocrinology & diabetes : official journal, German Society of Endocrinology [and] German Diabetes Association (2014-06-19)
T Çavusoglu, O Erbas, T Karadeniz, O Akdemir, E Acikgoz, M Karadeniz, M I Tuglu, U Ates
ABSTRACT
One of the major concerns is a nephropathy in diabetes, which applies many different kinds of medicines. However, required level of the treatment of renal disease has not been achieved. To investigate and compare the effect of the enalapril and the exenatide on diabetic nephropathy in rats developed diabetes by streptozosin. 32 male Sprague Dawley rats were divided into 4 groups: (1) Control, (2) Diabetic (DM), (3) DM+ Enalapril, and (4) DM+ exenatide groups. Then, the animals were euthanized and their blood samples were collected by cardiac puncture for blood glucose; blood urea nitrogen (BUN), creatinin, and nephrectomy were performed for histopathologic examination, and urine samples were taken on stick for proteinuria. Administration of the enalapril or the exenatide in diabetic rats resulted in a significant reduction both fibronectin, induced nitric oxide synthase (i-NOS) expression in glomerular area and urine protein levels. It was shown that both of enalapril and exenatide protected the renal glomerulus more than diabetic group in the nephropathy histopathologically. The beneficial effects of enalapril and exenatide which reduces fibronectin, i-NOS expression and urine protein levels or increases recovery of glomerules, might be used for preventing the harmful effects of diabetic nephropathy.
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O