  • Search Results
  • Neuronal and non-neuronal bradykinin receptors are involved in the contraction and/or relaxation to the pig bladder neck smooth muscle.
Neuronal and non-neuronal bradykinin receptors are involved in the contraction and/or relaxation to the pig bladder neck smooth muscle.
Neurourology and urodynamics (2013-07-13)
Ana S F Ribeiro, Vítor S Fernandes, María Pilar Martínez, Ana Martínez-Sáenz, María Ruth Pazos, Luis M Orensanz, Paz Recio, Salvador Bustamante, Joaquín Carballido, Albino García-Sacristán, Dolores Prieto, Medardo Hernández
ABSTRACT
The current study investigates the role played by bradykinin (BK) receptors in the contractility to the pig bladder neck smooth muscle. Bladder neck strips were mounted in myographs for isometric force recordings and BK receptors expression was also determined by immunohistochemistry. B2 receptor expression was observed in the muscular layer and urothelium whereas B1 expression was consistent detected in urothelium. A strong B2 immunoreactivity was also observed within nerve fibers among smooth muscle bundles. On urothelium-denuded preparations basal tone, BK induced concentration-dependent contractions which were reduced in urothelium-intact samples, by extracellular Ca(2+) removal and by blockade of B2 receptors and voltage-gated Ca(2+) (VOC) and non-VOC channels, and increased by cyclooxygenase (COX) inhibition. On phenylephrine-precontracted denuded strips, under non-adrenergic non-cholinergic (NANC) conditions, electrical field stimulation-elicited frequency-dependent relaxations which were reduced by B2 receptor blockade. In urothelium-intact samples, the B1 receptor agonist kallidin promoted concentration-dependent relaxations which were reduced by blockade of B1 receptors, COX, COX-1 and large-conductance Ca(2+) -activated K(+) (BKCa ) channels and abolished in urothelium-denuded samples and in K(+) -enriched physiological saline solution-precontracted strips. These results suggest that BK produces contraction of pig bladder neck via smooth muscle B2 receptors coupled to extracellular Ca(2+) entry via VOC and non-VOC channels with a minor role for intracellular Ca(2+) mobilization. Facilitatory neuronal B2 receptors modulating NANC inhibitory neurotransmission and urothelial B1 receptors producing relaxation via the COX-1 pathway and BKCa channel opening are also demonstrated. Neurourol. Urodynam. 33:558-565, 2014. © 2013 Wiley Periodicals, Inc.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Atropine, ≥99% (TLC), powder
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Supelco
Nifedipine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Atropine, European Pharmacopoeia (EP) Reference Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
3-(Trifluoromethyl)pyrazole, 99%
USP
Nifedipine, United States Pharmacopeia (USP) Reference Standard
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-MS4A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-BBS9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Supelco
Atropine, analytical standard
Nifedipine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-UGT1A1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-UGT1A1 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-MS4A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution