MilliporeSigma
Testicular synthesis and vitamin D action.
The Journal of clinical endocrinology and metabolism (2014-06-18)
D Hofer, J Münzker, V Schwetz, M Ulbing, K Hutz, Ph Stiegler, R Zigeuner, T R Pieber, H Müller, B Obermayer-Pietsch
ABSTRACT
The vitamin D system has pleiotropic effects not only in bone metabolism. Its role in testicular steroidogenesis is new and deserves intensive research. We hypothesize that vitamin D, especially 1,25 dihydroxyvitamin D3 [1,25(OH)2D3 (calcitriol)] induces male steroidogenesis and intend to identify its impact on genes and pathways in testicular androgen regulation. Human adult primary testicular cells were isolated, treated with 1,25(OH)2D3, and their gene expression levels profiled by microarray analysis. Highly regulated genes were confirmed by real-time quantitative PCR. In addition, the effects of 1,25(OH)2D3 in combination with LH and IGF-I on the gene expression level of androgens were assessed. T levels in the culture media were determined by a high-resolution ELISA. The expression of vitamin D receptor was confirmed at baseline and after 1,25(OH)2D3 stimulation using immunocytochemistry. Microarrays depicted 63 genes significantly regulated by 1,25(OH)2D3, including genes related to male androgen and vitamin D metabolism, mainly triggered by the vitamin D receptor/retinoid X receptor activation. 1,25(OH)2D3 led to significant changes in the expression profiles of reproductive genes and significantly increased T synthesis in human testicular cell cultures. Data from our human primary testicular cell culture model suggest that vitamin D plays a major role in male steroidogenesis in vitro.
Product Description
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
trans-Androsterone
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Adenine, European Pharmacopoeia (EP) Reference Standard
SAFC
HEPES
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)