  • Search Results
  • Effects of surface displayed targeting ligand GE11 on liposome distribution and extravasation in tumor.
Effects of surface displayed targeting ligand GE11 on liposome distribution and extravasation in tumor.
Molecular pharmaceutics (2014-09-03)
Hailing Tang, Xiaojing Chen, Mengjie Rui, Wenqiang Sun, Jian Chen, Jinliang Peng, Yuhong Xu
ABSTRACT
Targeting ligands displayed on liposome surface had been used to mediate specific interactions and drug delivery to target cells. However, they also affect liposome distribution in vivo, as well as the tissue extravasation processes after IV injection. In this study, we incorporated an EGFR targeting peptide GE11 on liposome surfaces in addition to PEG at different densities and evaluated their targeting properties and antitumor effects. We found that the densities of surface ligand and PEG were critical to target cell binding in vitro as well as pharmacokinetic profiles in vivo. The inclusion of GE11-PEG-DSPE and PEG-DSPE at 2% and 4% mol ratios in the liposome formulation mediated a rapid accumulation of liposomes within 1 h after IV injection in the tumor tissues surrounding neovascular structures. This is in addition to the EPR effect that was most prominently described for surface PEG modified liposomes. Therefore, despite the fact that the distribution of liposomes into interior tumor tissues was still limited by diffusion, GE11 targeted doxorubicin loaded liposomes showed significantly better antitumor activity in tumor bearing mice as a result of the fast active-targeting efficiency. We anticipate these understandings can benefit further optimization of targeted drug delivery systems for improving efficacy in vivo.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-EGFR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-EGFR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, ≥97%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 102618, purified immunoglobulin, lyophilized powder
Sigma-Aldrich
Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 29.1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone F4, ascites fluid
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Monoclonal Anti-EGFR antibody produced in mouse, clone 4H2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-EGFR antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0815, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Rabbit anti-EGFR Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anti-EGFR (Ab-1197) antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody