  • Search Results
  • Differential associations between childhood trauma subtypes and adolescent HPA-axis functioning.
Differential associations between childhood trauma subtypes and adolescent HPA-axis functioning.
Psychoneuroendocrinology (2015-02-24)
Kate R Kuhlman, Elisa G Geiss, Ivan Vargas, Nestor L Lopez-Duran
ABSTRACT
Studies examining the association between childhood trauma exposure and neuroendocrine functioning have returned inconsistent findings. To date, few studies have accounted for the role exposure to different types of childhood trauma may have on different neuroendocrine adaptations, and no study has examined this association using multiple indices of hypothalamic-pituitary-adrenal axis (HPA-axis) functioning. The purpose of this study was to characterize the unique associations between exposure to physical abuse, emotional abuse, and non-intentional trauma, and multiple indices of HPA-axis functioning. A community sample of 138 youth (aged 9-16) completed the Socially Evaluated Cold Pressor Task (SE-CPT) while their parents completed the Early Trauma Inventory (ETI). All youth then collected 4 diurnal salivary cortisol samples at home across 2 consecutive weekdays. High reported exposure to non-intentional trauma was associated with intact diurnal regulation but elevated cortisol at bedtime, physical abuse was associated with faster reactivity to acute stress, and emotional abuse was associated with delayed recovery of cortisol following acute stress. Taken together, there was a heterogeneous relationship among different indices of HPA-axis functioning and trauma subtype. Different types of childhood trauma exposure are related to distinct anomalies in HPA-axis functioning. This study underscores the importance of research incorporating multiple indices of HPA-axis functioning to inform our understanding of the underlying neuroendocrine dysregulation that may later lead to stress-related psychopathology.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard