  • Search Results
  • Cisplatin in combination with Phenethyl Isothiocyanate (PEITC), a potential new therapeutic strategy for malignant pleural mesothelioma.
Cisplatin in combination with Phenethyl Isothiocyanate (PEITC), a potential new therapeutic strategy for malignant pleural mesothelioma.
Oncotarget (2014-11-02)
Iza Denis, Laurent Cellerin, Marc Gregoire, Christophe Blanquart
ABSTRACT
Malignant pleural mesothelioma (MPM) is a very aggressive form of cancer with a poor diagnosis and prognosis. The first line treatment for MPM is a combination of cisplatin and Pemetrexed, which displayed limited efficacy and severe side effects. The naturally occurring compound phenethyl isothiocyanate (PEITC) previously showed interesting anti-tumor properties on several cancer cell lines. We thus aim at evaluating PEITC used alone or in combination with cisplatin in order to improve MPM treatment. Nine MPM cell lines and primary mesothelial cells (PMC), co-cultured or not with M2 macrophages present in MPM microenvironment, were used to assess PEITC and cisplatin anti-tumor properties. Compounds were used alone or in combination. Both PEITC and cisplatin were cytotoxic on MPM cells in a dose dependent manner. We herein showed that PEITC-induced cytotoxicity was due to the generation of reactive oxygen species. Moreover, we showed that cisplatin-PEITC combination allowed the potentialization of both compounds' cytotoxic effects and prevented the emergence of resistant MPM cells. Interestingly, PMC were not sensitive to the combination. Finally, we showed that M2 macrophages did not alter the anti-tumor properties of the combination. Cisplatin-PEITC combination thus represents a promising strategy to induce a selective toxicity towards malignant cells.
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Supelco
Aphidicolin, analytical standard
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard