  • Search Results
  • Chiral chromatography-tandem mass spectrometry applied to the determination of pro-resolving lipid mediators.
Chiral chromatography-tandem mass spectrometry applied to the determination of pro-resolving lipid mediators.
Journal of chromatography. A (2014-08-16)
Julia Homann, Christoph Lehmann, Astrid S Kahnt, Dieter Steinhilber, Michael J Parnham, Gerd Geisslinger, Nerea Ferreirós
ABSTRACT
Pro-resolving lipid mediators are a class of endogenously synthesized molecules derived from different fatty acids, such as arachidonic, docosahexaenoic or eicosapentaenoic acid, which are derived into four different product families: lipoxins, resolvins, maresins and protectins. For quantitation of these compounds, a sensitive, selective and robust liquid chromatography-tandem mass spectrometry method was developed and validated for the simultaneous quantitation of lipoxin A4, 6-epi-lipoxin A4, lipoxin B4 and lipoxin A5, the D-series resolvins D1 and D2 as well as aspirin-triggered lipoxin A4 and resolvin D1, maresin and protectin and the pathway markers 17(S)-hydroxy-docosahexaenoic acid and 17(R)-hydroxy-docosahexaenoic acid in cell culture supernatants. For this purpose, a chiral column was connected in series with a reversed-phase column to achieve efficient analyte separation and high sensitivity. Sample pre-treatment included a fast and simple liquid-liquid extraction procedure. Limits of quantitation in the range of 0.1-0.5ng/mL cell culture media, absolute recoveries between 90 and 115%, intra- and interday precision of less than 13% and an accuracy of less than 11% were obtained. Stability of the samples after 60 days storage at -80°C, three freeze/thaw cycles and 4h at room temperature has been demonstrated for all analytes. Sample extracts can be stored at 7°C for 24h without degradation of the analytes. Deviations of less than 13% in the accuracy, evaluated in terms of relative error, were obtained. The suitability of the method has been demonstrated in cell culture supernatants of human polymorphonuclear leukocytes, stimulated with 15R-hydroxy-eicosatetraenoic acid and in cell culture media of human polymorphonuclear leukocytes co-incubated with human platelets. From all studied analytes, lipoxin A4 and 6-epi-lipoxin A4 were found in cell culture media under both incubation conditions, while 15-epi-lipoxin A4 was additionally detected in cell culture supernatants of polymorphonuclear leukocytes stimulated with 15R-hydroxy-eicosatetraenoic acid.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Calcium ionophore III, Selectophore, function tested
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Lipoxin A4, ethanol solution
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard