  • Search Results
  • The immunomodulatory effects of pegylated liposomal doxorubicin are amplified in BRCA1--deficient ovarian tumors and can be exploited to improve treatment response in a mouse model.
The immunomodulatory effects of pegylated liposomal doxorubicin are amplified in BRCA1--deficient ovarian tumors and can be exploited to improve treatment response in a mouse model.
Gynecologic oncology (2014-04-01)
Gina Mantia-Smaldone, Lukas Ronner, Anne Blair, Victoria Gamerman, Christopher Morse, Sandra Orsulic, Stephen Rubin, Phyllis Gimotty, Sarah Adams
ABSTRACT
Women with BRCA-associated ovarian cancer demonstrate excellent responses to Pegylated Liposomal Doxorubicin (PLD). PLD has also been shown to enhance T cell recognition of tumor cells. Here we characterize immunophenotypic changes associated with BRCA1 dysfunction in ovarian cancer cells, and evaluate the T cell contribution to the therapeutic efficacy of PLD in a BRCA1- ovarian cancer model to determine whether enhanced anti-tumor immunity contributes to the improved response to PLD in BRCA1- ovarian cancers. The immunophenotype of BRCA1- and wild-type (WT) ovarian cancer cells and their response to PLD were compared in vitro using flow cytometry. T cell recruitment to BRCA1- tumors was evaluated with flow cytometry and immunohistochemistry. The contribution of T cell populations to the therapeutic effect of PLD in a BRCA1- model was evaluated using immunodepleting antibodies with PLD in vivo. The cytotoxic response to PLD was similar in BRCA1- and WT cells in vitro. BRCA1- inactivation resulted in higher expression of Fas and MHC-I at baseline and after PLD exposure. PLD prolonged the survival of BRCA1- tumor bearing mice and increased intratumoral T cell recruitment. CD4+ depletion combined with PLD significantly prolonged overall survival (p=0.0204) in BRCA1- tumor-bearing mice. Differences in the immunophenotype of BRCA1- and WT cells are amplified by PLD exposure. The enhanced immunomodulatory effects of PLD in BRCA1- tumors may be exploited therapeutically by eliminating suppressive CD4+ T cells. Our results support further study of combination therapy using PLD and immune agents, particularly in women with BRCA gene mutations.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Monoclonal Anti-CD95 antibody produced in mouse, clone LT95, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-FAS antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-FAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-FAS (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-FAS (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-FAS antibody produced in mouse, clone 2G3, purified immunoglobulin, buffered aqueous solution
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC