  • Search Results
  • β-cypermethrin-induced acute neurotoxicity in the cerebral cortex of mice.
β-cypermethrin-induced acute neurotoxicity in the cerebral cortex of mice.
Drug and chemical toxicology (2014-03-22)
DeQing Cao, Nan Chen, ChunXiao Zhu, Yue Zhao, Li Liu, Jun Yang, Li An
ABSTRACT
A Type II pyrethroid pesticide β-cypermethrin is widely used in agriculture and domestic applications for pest control. However, the effect of β-cypermethrin on the glutamate neurotransmitter has not been well-documented. In the current study, mice were treated with 20, 40, or 80 mg/kg β-cypermethrin by a single oral gavage, with corn oil as a vehicle control. Four hours after treatment, we investigated glutamate levels and glutamate-metabolizing enzyme (phosphate-activated glutaminase, PAG; glutamine synthetase, GS) activities in the cerebral cortex of mice, using a HPLC system with ultraviolet detectors and a colorimetric assay. Glutamate uptake levels in the synaptosomes of cerebral cortex and mRNA expression levels of PAG, GS, and glutamate transporter-1 (GLT-1) in the cerebral cortex were detected by a radioactive labeling method and qRT-PCR, respectively. Toxic symptoms were observed in mice treated with 40 or 80 mg/kg β-cypermethrin. Compared with the control, significant decreases in glutamate level and GS activity, and an obvious increase in synaptosomal glutamate uptake, were found in the cerebral cortex of mice treated with 80 mg/kg β-cypermethrin. No significant changes were found among groups in PAG activity or PAG, GS, and GLT-1 mRNA expression levels. These results suggest that β-cypermethrin treatment may reduce the glutamate level in the mouse cerebral cortex, which is associated with decreased GS activity and increased synaptosomal glutamate uptake. Our findings provide a partial explanation for the neurotoxic effects of synthetic β-cypermethrin insecticides.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for amino acid labeling, powder and chunks, ≥99% (HPLC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dansyl chloride, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
Dansyl chloride, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC