MilliporeSigma
Ultrafast cooling reveals microsecond-scale biomolecular dynamics.
Nature communications (2014-12-18)
Mark E Polinkovsky, Yann Gambin, Priya R Banerjee, Michael J Erickstad, Alex Groisman, Ashok A Deniz
ABSTRACT
The temperature-jump technique, in which the sample is rapidly heated by a powerful laser pulse, has been widely used to probe the fast dynamics of folding of proteins and nucleic acids. However, the existing temperature-jump setups tend to involve sophisticated and expensive instrumentation, while providing only modest temperature changes of ~10-15 °C, and the temperature changes are only rapid for heating, but not cooling. Here we present a setup comprising a thermally conductive sapphire substrate with light-absorptive nano-coating, a microfluidic device and a rapidly switched moderate-power infrared laser with the laser beam focused on the nano-coating, enabling heating and cooling of aqueous solutions by ~50 °C on a 1-μs time scale. The setup is used to probe folding and unfolding dynamics of DNA hairpins after direct and inverse temperature jumps, revealing low-pass filter behaviour during periodic temperature variations.
Product Description
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt, ≥96%