  • Search Results
  • Lymphocytic mitochondrial aconitase activity is reduced in Alzheimer's disease and mild cognitive impairment.
Lymphocytic mitochondrial aconitase activity is reduced in Alzheimer's disease and mild cognitive impairment.
Journal of Alzheimer's disease : JAD (2014-10-18)
Francesca Mangialasche, Mauro Baglioni, Roberta Cecchetti, Miia Kivipelto, Carmelinda Ruggiero, Danilo Piobbico, Lothar Kussmaul, Roberto Monastero, Stefano Brancorsini, Patrizia Mecocci
ABSTRACT
Specific mechanisms behind the role of oxidative/nitrosative stress and mitochondrial dysfunction in Alzheimer's disease (AD) pathogenesis remain elusive. Mitochondrial aconitase (ACO2) is a Krebs cycle enzyme sensitive to free radical-mediated damage. We assessed activity and expression of ACO2 extracted from blood lymphocytes of subjects with AD, mild cognitive impairment (MCI), older adults with normal cognition (OCN, age ≥65 years), and younger adults with normal cognition (YCN, age <65 years). Plasma levels and activities of antioxidants were also measured. Blood samples were collected from 28 subjects with AD, 22 with MCI, 21 OCN, and 19 YCN. ACO2 activity was evaluated in a subsample before and after in vitro exposure to free radicals. ACO2 activity was significantly lower in AD and MCI cases than controls: ACO2 median activity was 0.64 ± 0.21 U/mg protein for AD, 0.93 ± 0.28 U/mg protein for MCI, 1.17 ± 0.78 U/mg protein for OCN subjects, and 1.23 ± 0.43 U/mg protein for YCN individuals. In subjects with AD and MCI, ACO2 expression was lower than OCN subjects, and ACO2 activity correlated with vitamin E plasma levels (rho: 0.64, p < 0.001) and Mini-Mental State Examination total score (rho: 0.82, p < 0.001). Furthermore, free radicals exposure reduced ACO2 activity more in individuals with AD than in OCN subjects. Our results suggest that ACO2 activity is reduced in peripheral lymphocytes of subjects with AD and MCI and correlates with antioxidant protection. Further studies are warranted to verify the role of ACO2 in AD pathogenesis and its importance as a marker of AD progression.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)