  • Search Results
  • Hyperoside regulates the level of thymic stromal lymphopoietin through intracellular calcium signalling.
Hyperoside regulates the level of thymic stromal lymphopoietin through intracellular calcium signalling.
Phytotherapy research : PTR (2013-12-18)
Na-Ra Han, Ji-Hyun Go, Hyung-Min Kim, Hyun-Ja Jeong
ABSTRACT
Hyperoside (HYP) is the principle active component of Crataegus pinnatifida. Thymic stromal lymphopoietin (TSLP) plays a vital role in the pathogenesis of allergic reactions. Here, we investigated how HYP regulates the levels of TSLP in a human mast cell line, HMC-1 cells. We analyzed the levels of TSLP by treatment with HYP in phorbol myristate acetate plus calcium ionophore A23187-stimulated HMC-1 cells with ELISA and a polymerase chain reaction analysis. We also analyzed the pathway that HYP regulates TSLP by measuring the level of fluorescent intracellular calcium and using a Western blot analysis. HYP decreased the level of intracellular calcium in stimulated HMC-1 cells. It also significantly decreased the production and mRNA expression of TSLP in stimulated HMC-1 cells. It significantly decreased the levels of receptor-interacting protein 2 and active caspase-1 in stimulated HMC-1 cells. HYP significantly decreased the translocation of NF-κB into the nucleus and degradation of IκBα in the cytoplasm in stimulated HMC-1 cells. Furthermore, it significantly decreased the production and mRNA expression of interleukin-1β and interleukin-6 in stimulated HMC-1 cells. Taken together, our findings establish HYP as a potential agent for the treatment of allergic reactions.
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-6 sR ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
1,2-Bis(2-amino-5-methylphenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, ≥95% (HPLC)
Sigma-Aldrich
Caspase 1 human, recombinant, expressed in E. coli, ≥5000 units/mg protein
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Human IL-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate
Sigma-Aldrich
Human IL-6 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant