MilliporeSigma
Temporo-spatial requirement of Smad4 in dentin formation.
Biochemical and biophysical research communications (2015-03-17)
Tak-Heun Kim, Cheol-Hyeon Bae, Ju-Yeon Lee, Jeong-Chae Lee, Seung-O Ko, Yang Chai, Eui-Sic Cho
ABSTRACT
The TGF-β/BMP family plays an important role in multiple stages of tooth development. TGF-β/BMP signaling is required for odontoblast differentiation and dentin formation; however, the precise molecular mechanisms underlying dentin formation remain unclear. To address the role of TGF-β/BMP signaling in dentin formation, we analyzed mice in which Smad4, a key intracellular mediator of TGF-β/BMP signaling, was subjected to tissue-specific ablation under the control of Dspp, OC, or Col1a1 promoters. Three independent Smad4 conditional knockout mice exhibited various dentin defects in the crowns and roots of their molars depending on the transactivator. In all mutant molars, crown dentin thickness was thinner than that of the control. In addition, impaired dentin was found in the cervical region and root furcation area. Although the initial differentiation of odontoblasts was normal, odontoblast polarity abruptly decreased and the expression of Col1a1, OC, and Dspp was reduced in the odontoblasts of mutant molars. In Dspp-Cre-mediated Smad4 disruption mice, primary dentin formation was slightly delayed, while secondary dentin formation was severely affected in the cervical region of the molars. These results indicate that TGF-β/BMP signaling is required for odontoblast maturation and dentin formation in a stage- and site-dependent manner.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer