  • Search Results
  • Does an injection of a stromal vascular fraction containing adipose-derived mesenchymal stem cells influence the outcomes of marrow stimulation in osteochondral lesions of the talus? A clinical and magnetic resonance imaging study.
Does an injection of a stromal vascular fraction containing adipose-derived mesenchymal stem cells influence the outcomes of marrow stimulation in osteochondral lesions of the talus? A clinical and magnetic resonance imaging study.
The American journal of sports medicine (2014-08-12)
Yong Sang Kim, Ho Jin Lee, Yun Jin Choi, Yong Il Kim, Yong Gon Koh
ABSTRACT
Marrow stimulation for the treatment of osteochondral lesions of the talus (OLTs) is controversial in patients with poor prognostic factors of OLTs. Currently, mesenchymal stem cells (MSCs) are expected to biologically augment the treatment of OLTs. To compare the clinical and magnetic resonance imaging (MRI) outcomes between an injection of MSCs with marrow stimulation and marrow stimulation alone in patients with OLTs. Cohort study; Level of evidence, 3. A total of 49 patients (50 ankles) with OLTs underwent follow-up MRI after arthroscopic treatment. Among these 50 ankles, 26 underwent marrow stimulation alone (conventional group), and 24 underwent marrow stimulation with an injection of a stromal vascular fraction (SVF) containing MSCs (MSC group). Clinical outcomes were evaluated according to the visual analog scale (VAS) for pain, American Orthopaedic Foot and Ankle Society (AOFAS) Ankle-Hindfoot Scale, and Tegner activity scale. The magnetic resonance observation of cartilage repair tissue (MOCART) score was used for the MRI evaluation of repaired lesions. The mean VAS score, AOFAS score, and Tegner score improved from 7.1 ± 1.2, 68.5 ± 5.6, and 3.4 ± 0.6 to 3.9 ± 0.8, 78.3 ± 4.9, and 3.5 ± 0.8, respectively, in the conventional group and from 7.1 ± 0.8, 67.7 ± 4.7, and 3.4 ± 0.5 to 3.2 ± 0.8, 83.3 ± 7.0, and 3.9 ± 0.7, respectively, in the MSC group. All clinical outcomes, including the VAS, AOFAS, and Tegner scores, improved significantly in the MSC group compared with the conventional group (P = .003, .009, and .041, respectively). There was a significant difference (P = .037) in the mean MOCART score between the conventional and MSC groups (49.4 ± 16.6 vs 62.1 ± 21.8, respectively), and significant correlations of the MOCART score with clinical outcomes were found in both groups (P < .05). Patient age (≥46.1 years), large lesion size (≥151.2 mm(2)), and the presence of subchondral cysts were associated with a worse MOCART score in the conventional group (P = .015, .004, and .013, respectively) but not in the MSC group. Clinical and MRI outcomes of an injection of an SVF containing MSCs with marrow stimulation were encouraging, compared with marrow stimulation alone, for the treatment of OLTs. Therefore, an injection of an SVF containing MSCs with marrow stimulation should be considered as a treatment for OLTs, even when poor prognostic factors, including older age, large-sized lesion, or the presence of subchondral cysts, exist.
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)