  • Search Results
  • Biosynthesis of lactosylfructoside by an intracellular levansucrase from Bacillus methylotrophicus SK 21.002.
Biosynthesis of lactosylfructoside by an intracellular levansucrase from Bacillus methylotrophicus SK 21.002.
Carbohydrate research (2014-12-17)
Chao Wu, Tao Zhang, Wanmeng Mu, Ming Miao, Bo Jiang
ABSTRACT
Lactosylfructoside is a functional oligosaccharide consisting of D-glucose, D-galactose, and D-fructose. In this work, lactosylfructoside was biosynthesized from sucrose as a fructosyl donor and lactose as an acceptor by an intracellular levansucrase derived from strain Bacillus methylotrophicus (B. methylotrophicus) SK21.002. The trisaccharide was purified from the product using a high performance liquid chromatography (HPLC) system and was confirmed to be lactosylfructoside by nuclear magnetic resonance (NMR) spectroscopy. The biosynthesis conditions (such as pH, temperature, enzyme dosage, substrate concentrations, and the concentration ratio of the two substrates) for lactosylfructoside production were optimized. The optimum conditions for lactosylfructoside preparation were a pH of 6.5, temperature of 37 °C, and enzyme dosage of 8 U/g substrates. The concentration of substrates (total lactose and sucrose) was 400mg/mL, and the ratio of lactose to sucrose was 1:1. The optimum time for lactosylfructoside production was 20 h, the yield of lactosylfructoside under the optimal conditions was 143 mg/mL, and the lactosylfructoside conversion efficiency was 36%.
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", 99.96 atom % D
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)