  • Search Results
  • Plasma, salivary and urinary cortisol levels following physiological and stress doses of hydrocortisone in normal volunteers.
Plasma, salivary and urinary cortisol levels following physiological and stress doses of hydrocortisone in normal volunteers.
BMC endocrine disorders (2014-11-27)
Caroline Jung, Santo Greco, Hanh H T Nguyen, Jui T Ho, John G Lewis, David J Torpy, Warrick J Inder
ABSTRACT
Glucocorticoid replacement is essential in patients with primary and secondary adrenal insufficiency, but many patients remain on higher than recommended dose regimens. There is no uniformly accepted method to monitor the dose in individual patients. We have compared cortisol concentrations in plasma, saliva and urine achieved following "physiological" and "stress" doses of hydrocortisone as potential methods for monitoring glucocorticoid replacement. Cortisol profiles were measured in plasma, saliva and urine following "physiological" (20 mg oral) or "stress" (50 mg intravenous) doses of hydrocortisone in dexamethasone-suppressed healthy subjects (8 in each group), compared to endogenous cortisol levels (12 subjects). Total plasma cortisol was measured half-hourly, and salivary cortisol and urinary cortisol:creatinine ratio were measured hourly from time 0 (between 0830 and 0900) to 5 h. Endogenous plasma corticosteroid-binding globulin (CBG) levels were measured at time 0 and 5 h, and hourly from time 0 to 5 h following administration of oral or intravenous hydrocortisone. Plasma free cortisol was calculated using Coolens' equation. Plasma, salivary and urine cortisol at 2 h after oral hydrocortisone gave a good indication of peak cortisol concentrations, which were uniformly supraphysiological. Intravenous hydrocortisone administration achieved very high 30 minute cortisol concentrations. Total plasma cortisol correlated significantly with both saliva and urine cortisol after oral and intravenous hydrocortisone (P <0.0001, correlation coefficient between 0.61 and 0.94). There was no difference in CBG levels across the sampling period. An oral dose of hydrocortisone 20 mg is supraphysiological for routine maintenance, while stress doses above 50 mg 6-hourly would rarely be necessary in managing acute illness. Salivary cortisol and urinary cortisol:creatinine ratio may provide useful alternatives to plasma cortisol measurements to monitor replacement doses in hypoadrenal patients.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Hydrocortisone, VETRANAL®, analytical standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard