  • Search Results
  • PEDF improves mitochondrial function in RPE cells during oxidative stress.
PEDF improves mitochondrial function in RPE cells during oxidative stress.
Investigative ophthalmology & visual science (2014-09-13)
Yuan He, Kar Wah Leung, Yuan Ren, Jinzhi Pei, Jian Ge, Joyce Tombran-Tink
ABSTRACT
Oxidative stress plays an important role in health and aging. We have shown that oxidative stress impairs mitochondrial function and promotes RPE cell death in an age-dependent manner. This study investigates the role of pigment epithelium-derived factor (PEDF) in limiting oxidative stress-induced damage to RPE cells through mitochondrial pathways. Three groups of early-passaged RPE cells from donors 50 to 55, 60 to 65, and 70 to 75 years old (yo) were either preconditioned with PEDF followed by exposure to sublethal doses of hydrogen peroxide (H2O2) or post-treated with PEDF after H2O2 treatment. Effects of PEDF on mitochondrial function and cell viability were examined. Oxidative stress induced an age-dependent increase in LDH release, reactive oxygen species (ROS) levels, and cell death and a decrease in adenosine triphosphate (ATP) production and mitochondrial membrane potential (ΔΨm) in human RPE cells. Preconditioning or poststressed treatment with PEDF resulted in increased cell viability, inhibition of cytochrome c release and caspase 3 cleavage, and improved mitochondria function denoted by a decrease in ROS generation and increases in ATP production and ΔΨm. Oxidative stress also disrupted the reticular network, trafficking, and distribution of the mitochondria and blocked activation of phosphatidylinositol 3 kinase (PI3K), Akt, and Erk signaling in the cells. These effects were more pronounced in RPE cells from individuals>60 yo compared to the 50 to 55 yo age group. Pigment epithelium-derived factor mitigated negative effects of oxidative stress on mitochondrial remodeling and cellular distribution and unblocked its control of PI3K/Akt and mitogen-activated protein kinase (MAPK) signaling. Although PEDF potentiated both PI3K/Akt and MAPK signaling in the cells, stabilization of mitochondrial networks and function was dependent on its activation of PI3K/Akt. Specificity of PEDF's activity was confirmed using the pharmacological inhibitors LY294002, SH6, and U0126. We also show that in the absence of oxidative stress, pharmacological inhibition of the PI3K/Akt pathway alone was sufficient to disrupt mitochondrial structure and function. In addition, PEDF blocked effects of oxidative stress on expression of cyclophilin D and UCP2, genes controlling mitochondrial function, and the apoptotic genes caspase 3, Bax, and Bcl2. Control of ROS levels by PEDF was specifically linked to UCP2 regulation since PEDF-induced expression of this gene in UCP2-deficient cells was associated with a decrease in ROS production. We provide evidence that PEDF promotes resilience of aging RPE cells to oxidative stress by stabilizing mitochondrial networks and function and that mitochondrial dynamics in human RPE cells are controlled, in part, through the PI3K/Akt pathway.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
JC-1, solid
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%